Anti-CDK9 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9941
10707-340EA 786.15 USD
10707-340
Anti-CDK9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CDK9.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:3000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: CDK9
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR