Anti-CIITA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9942
10707-986EA 786.15 USD
10707-986
Anti-CIITA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant CIITA.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CIITA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR