Anti-MAP3K5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9947
10677-704EA 786.15 USD
10677-704
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphpeptide of MAP3K5.

Recommended Dilutions: ELISA: 1:5000-1:10000; Western Blot: 1:1000

Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR