Anti-MYL6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9961
10675-010EA 786.15 USD
10675-010
Anti-MYL6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MYL6.

Recommended Dilutions: ELISA: 1:120000-1:135000; Western Blot: 1:1000-1:5000; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: MYL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR