Anti-AP3D1 Goat Polyclonal Antibody

Supplier: Abnova
PAB9965
10689-512EA 786.15 USD
10689-512
Anti-AP3D1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of AP3D1.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: AP3D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR