Anti-CCR3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26095
10683-476EA 515 USD
10683-476
Anti-CCR3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of CCR3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 8 ?g/ml

Type: Primary
Antigen: CCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR