Anti-GPR17 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26290
10684-452EA 515 USD
10684-452
Anti-GPR17 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of GPR17.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2 ?g/ml

Type: Primary
Antigen: GPR17
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR