Anti-GPR151 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26323
10684-382EA 653.85 USD
10684-382
Anti-GPR151 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GPR151.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-5 ?g/ml

Type: Primary
Antigen: GPR151
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR