Anti-GPR3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26275
10684-586EA 653.85 USD
10684-586
Anti-GPR3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GPR3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 6 ?g/ml

Type: Primary
Antigen: GPR3
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Gorilla, Hamster, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR