Anti-PTGDR2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26316
10684-604EA 653.85 USD
10684-604
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GPR44.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-6 ?g/ml

Type: Primary
Antigen: PTGDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR