Anti-GPR35 Rabbit Polyclonal Antibody

PAB26276
10691-838EA 647.69 USD
10691-838
Anti-GPR35 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of GPR35.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 9-22 ?g/ml

Type: Primary
Antigen: GPR35
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR