Anti-GALR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB26208
10684-144EA 515 USD
10684-144
Anti-GALR1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of GALR1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 19 ?g/ml

Type: Primary
Antigen: GALR1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR