Anti-ATOH7 Mouse Monoclonal Antibody [clone: 1A5]

Supplier: Abnova
H00220202-M01
10581-056EA 335 USD
10581-056
Anti-ATOH7 Mouse Monoclonal Antibody [clone: 1A5]
Antibodies
Type: Primary
Antigen: ATOH7
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR