Anti-CCDC83 Mouse Monoclonal Antibody [clone: 3000]

Supplier: Abnova
H00220047-M01
10593-362EA 335 USD
10593-362
Anti-CCDC83 Mouse Monoclonal Antibody [clone: 3000]
Antibodies
Type: Primary
Antigen: CCDC83
Clonality: Monoclonal
Clone: 3000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR