![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ENKUR |
Antigen Name | enkurin, TRPC channel interacting protein |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | CFAP106, C10orf63 |
Gene ID | 219670 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |