Anti-HIPK1 Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Abnova
H00204851-M04
10588-824EA 335 USD
10588-824
Anti-HIPK1 Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
Type: Primary
Antigen: HIPK1
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR