Anti-NLRP11 Mouse Polyclonal Antibody

H00204801-B01
10565-234EA 310.77 USD
10565-234
Anti-NLRP11 Mouse Polyclonal Antibody
Type: Primary
Antigen: NLRP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR