Anti-STK32A Mouse Monoclonal Antibody [clone: 1C5]

Supplier: Abnova
H00202374-M02
10602-730EA 335 USD
10602-730
Anti-STK32A Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
Type: Primary
Antigen: STK32A
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR