Anti-STK32A Mouse Monoclonal Antibody [clone: 3E8-1D3]

Supplier: Abnova
H00202374-M01
10593-340EA 335 USD
10593-340
Anti-STK32A Mouse Monoclonal Antibody [clone: 3E8-1D3]
Antibodies
Type: Primary
Antigen: STK32A
Clonality: Monoclonal
Clone: 3E8-1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR