Anti-STK32A Rabbit Polyclonal Antibody

Supplier: Abnova
H00202374-D01
10602-736EA 376 USD
10602-736
Anti-STK32A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STK32A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR