Anti-ZSCAN4 Mouse Monoclonal Antibody [clone: 2C7]

Supplier: Abnova
H00201516-M04
10606-628EA 335 USD
10606-628
Anti-ZSCAN4 Mouse Monoclonal Antibody [clone: 2C7]
Antibodies
Type: Primary
Antigen: ZSCAN4
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR