Anti-CENPV Mouse Monoclonal Antibody [clone: 1D9]

Supplier: Abnova
H00201161-M05
10598-256EA 335 USD
10598-256
Anti-CENPV Mouse Monoclonal Antibody [clone: 1D9]
Antibodies
Type: Primary
Antigen: CENPV
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR