Anti-CENPV Mouse Polyclonal Antibody

H00201161-B01P
10568-476EA 316.92 USD
10568-476
Anti-CENPV Mouse Polyclonal Antibody
Type: Primary
Antigen: CENPV
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR