Anti-SPRED2 Mouse Monoclonal Antibody [clone: 6G8]

Supplier: Abnova
H00200734-M01
10602-010EA 335 USD
10602-010
Anti-SPRED2 Mouse Monoclonal Antibody [clone: 6G8]
Antibodies
Type: Primary
Antigen: SPRED2
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR