Anti-APOBEC3F Mouse Polyclonal Antibody

Supplier: Abnova
H00200316-A01
10559-122EA 243 USD
10559-122
Anti-APOBEC3F Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOBEC3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR