Anti-U2AF1L4 Mouse Polyclonal Antibody

Supplier: Abnova
H00199746-B01
10573-266EA 335 USD
10573-266
Anti-U2AF1L4 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: U2AF1L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR