![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | LDHD |
Antigen Name | Lactate dehydrogenase D |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | Lactate dehydrogenase D, Probable D-lactate dehydrogenase, DLD, LDHD, mitochondrial |
Gene ID | 197257 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |