Anti-AMN1 Mouse Polyclonal Antibody

H00196394-B01P
10563-168EA 316.92 USD
10563-168
Anti-AMN1 Mouse Polyclonal Antibody
Type: Primary
Antigen: AMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR