![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | NUDT10 |
Antigen Name | Nudix-type Motif 10 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | DIPP3-alpha, DIPP3a, APS2, Nudix (nucleoside diphosphate linked moiety X)-type motif 10 |
Gene ID | 170685 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |