Anti-ARX Mouse Monoclonal Antibody [clone: 4D12]

Supplier: Abnova
H00170302-M02
10577-338EA 335 USD
10577-338
Anti-ARX Mouse Monoclonal Antibody [clone: 4D12]
Antibodies
Type: Primary
Antigen: ARX
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR