![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ARX |
Antigen Name | Aristaless Related Homeobox |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 1G2 |
Host | Mouse |
ELISA | Yes |
Antigen Synonyms | MRX33, CT121, MRX29, MRX36, MRX38, PRTS, MRX43, MRXS1, MRX32, MRX54, MRX76, EIEE1, ISSX, MRX87 |
Gene ID | 170302 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |