Anti-GIMAP7 Mouse Polyclonal Antibody

Supplier: Abnova
H00168537-B01P
10560-882EA 335 USD
10560-882
Anti-GIMAP7 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GIMAP7 (GTPase, IMAP family member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR