Anti-RNF133 Mouse Monoclonal Antibody [clone: 3D6]

Supplier: Abnova
H00168433-M01
10599-896EA 335 USD
10599-896
Anti-RNF133 Mouse Monoclonal Antibody [clone: 3D6]
Antibodies
Type: Primary
Antigen: RNF133
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR