Anti-RNF133 Rabbit Polyclonal Antibody

Supplier: Abnova
H00168433-D01
10599-898EA 376 USD
10599-898
Anti-RNF133 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF133
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR