Anti-RNF133 Mouse Polyclonal Antibody

Supplier: Abnova
H00168433-B01P
10570-008EA 335 USD
10570-008
Anti-RNF133 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF133
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR