Anti-PRSS35 Mouse Polyclonal Antibody

H00167681-B01P
10568-692EA 316.92 USD
10568-692
Anti-PRSS35 Mouse Polyclonal Antibody
Type: Primary
Antigen: PRSS35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR