Anti-UGT3A2 Mouse Polyclonal Antibody

Supplier: Abnova
H00167127-B01
10573-450EA 335 USD
10573-450
Anti-UGT3A2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UGT3A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR