Anti-BBS12 Mouse Polyclonal Antibody

H00166379-B01P
10552-136EA 316.92 USD
10552-136
Anti-BBS12 Mouse Polyclonal Antibody
Type: Primary
Antigen: BBS12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR