Anti-OXER1 Mouse Polyclonal Antibody

H00165140-B01P
10566-466EA 316.92 USD
10566-466
Anti-OXER1 Mouse Polyclonal Antibody
Type: Primary
Antigen: OXER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR