Anti-CCDC138 Mouse Polyclonal Antibody

Supplier: Abnova
H00165055-B01
10556-810EA 335 USD
10556-810
Anti-CCDC138 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCDC138
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR