![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | DYX1C1 |
Antigen Name | Dyslexia susceptibility 1 candidate 1 |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | RD, DYX1, DYXC1, EKN1, DNAAF4, CILD25 |
Gene ID | 161582 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |