Anti-LDHAL6A Mouse Polyclonal Antibody

Supplier: Abnova
H00160287-B01
10562-892EA 335 USD
10562-892
Anti-LDHAL6A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LDHAL6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR