Anti-RASEF Mouse Monoclonal Antibody [clone: 1000]

Supplier: Abnova
H00158158-M02
10599-230EA 335 USD
10599-230
Anti-RASEF Mouse Monoclonal Antibody [clone: 1000]
Antibodies
Type: Primary
Antigen: RASEF
Clonality: Monoclonal
Clone: 1000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR