Anti-CLVS1 Mouse Polyclonal Antibody

H00157807-B01
10564-162EA 310.77 USD
10564-162
Anti-CLVS1 Mouse Polyclonal Antibody
Type: Primary
Antigen: CLVS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR