Anti-SH3RF2 Mouse Polyclonal Antibody

Supplier: Abnova
H00153769-A01
10571-388EA 235 USD
10571-388
Anti-SH3RF2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SH3RF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR