Anti-SGOL1 Mouse Monoclonal Antibody [clone: 3C11]

Supplier: Abnova
H00151648-M01
10601-258EA 335 USD
10601-258
Anti-SGOL1 Mouse Monoclonal Antibody [clone: 3C11]
Antibodies
Type: Primary
Antigen: SGOL1
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR