![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SGOL1 |
Antigen Name | Shugoshin-like 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
ImmunoPrecipitation | Yes |
Antigen Synonyms | Sgo1, SGO, CAID, NY-BR-85 |
Gene ID | 151648 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |