Anti-GDF7 Mouse Monoclonal Antibody [clone: 2G12]

Supplier: Abnova
H00151449-M06
10587-274EA 335 USD
10587-274
Anti-GDF7 Mouse Monoclonal Antibody [clone: 2G12]
Antibodies
Type: Primary
Antigen: GDF7 (growth differentiation factor 7)
Clonality: Monoclonal
Clone: 2G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR