Anti-GDF7 Mouse Monoclonal Antibody [clone: 3C2]

Supplier: Abnova
H00151449-M04
10587-286EA 335 USD
10587-286
Anti-GDF7 Mouse Monoclonal Antibody [clone: 3C2]
Antibodies
Type: Primary
Antigen: GDF7 (growth differentiation factor 7)
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR