Anti-FAM84A Mouse Monoclonal Antibody [clone: 1C2]

Supplier: Abnova
H00151354-M01
10595-444EA 335 USD
10595-444
Anti-FAM84A Mouse Monoclonal Antibody [clone: 1C2]
Antibodies
Type: Primary
Antigen: FAM84A (family with sequence similarity 84, member A)
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR